accessibility menu, dialog, popup

UserName

CAS RN 193202-37-8

H3CSOONOOOHClClCH3

CAS RN 193202-37-8

IUPAC Name: 4,5-dichloro-2-{10,10-dimethyl-3,3-dioxo-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-4-carbonyl}benzoic acid
Synonyms: 4,5-dichloro-2-{10,10-dimethyl-3,3-dioxo-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-4-carbonyl}benzoic acid
Molecular Weight (g/mol): 432.31
Molecular Formula: C18H19Cl2NO5S
InChi Key: RFABDSDXSYNRJG-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC11CS(=O)(=O)N(C1C2)C(=O)C1=CC(Cl)=C(Cl)C=C1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results