accessibility menu, dialog, popup

UserName

CAS RN 191732-72-6

IUPAC Name: 3-(4-amino-1-oxo-2,3-dihydro-1H-isoindol-2-yl)piperidine-2,6-dione
Synonyms: lenalidomide revlimid
Molecular Weight (g/mol): 259.27
Molecular Formula: C13H13N3O3
InChi Key: GOTYRUGSSMKFNF-UHFFFAOYNA-N
SMILES: NC1=CC=CC2=C1CN(C1CCC(=O)NC1=O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results