accessibility menu, dialog, popup

UserName

CAS RN 191-68-4

IUPAC Name: hexacyclo[12.12.0.0²,⁷.0⁸,¹³.0¹⁵,²⁰.0²¹,²⁶]hexacosa-1(14),2,4,6,8,10,12,15,17,19,21,23,25-tridecaene
Synonyms: dibenzo[g,p]chrysene
Molecular Weight (g/mol): 328.41
Molecular Formula: C26H16
InChi Key: GQDKQZAEQBGVBS-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=C2C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results