missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoronitrobenzene, 98%, Thermo Scientific™

Catalog No. AC397340050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397340050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397340050 Supplier Thermo Scientific Chemicals Supplier No. 397340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Density 1.5030g/mL
Boiling Point 91.0°C to 92.0°C (11.0mmHg)
Flash Point 88°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3NO2
Quantity 5g
Specific Gravity 1.503
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 2, 6-Difluoronitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 242-793-8

RUO – Research Use Only