accessibility menu, dialog, popup

UserName

CAS RN 1895-39-2

IUPAC Name: sodium 2-chloro-2,2-difluoroacetate
Synonyms: sodium 2-chloro-2,2-difluoroacetate
Molecular Weight (g/mol): 152.46
Molecular Formula: C2ClF2NaO2
InChi Key: MRTAVLDNYYEJHK-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(F)(F)Cl
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)