missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorodifluoroacetate 99.0+%, TCI America™

Catalog No. C0991500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0991500G 500 g
C099125G 25 g
C0991100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0991500G Supplier TCI America Supplier No. C0991500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Molecular Weight (g/mol) 152.457
MDL Number MFCD00064771
InChI Key MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonym sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Cl)[O-].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 152.46
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Chlorodifluoroacetate