accessibility menu, dialog, popup

UserName

CAS RN 187344-92-9

IUPAC Name: bis(3,5-dimethylphenyl)(oxo)-λ⁵-phosphanylium
Synonyms: bis(3,5-dimethylphenyl)(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 257.29
Molecular Formula: C16H18OP
InChi Key: LMXRTXPFJNGAAX-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)[P+](=O)C1=CC(C)=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results