accessibility menu, dialog, popup

UserName

H3COOCH3NSOOH3CCH3CH3

CAS RN 187324-66-9

IUPAC Name: 2-(N-benzyl2,4,6-trimethylbenzenesulfonamido)-1-phenylpropyl propanoate
Synonyms: 2-(N-benzyl2,4,6-trimethylbenzenesulfonamido)-1-phenylpropyl propanoate
Molecular Weight (g/mol): 479.64
Molecular Formula: C28H33NO4S
InChi Key: WNCDSLPLRUHTTL-UHFFFAOYNA-N
SMILES: CCC(=O)OC(C(C)N(CC1=CC=CC=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results