accessibility menu, dialog, popup

UserName

ClH2CCl

CAS RN 1871-57-4

IUPAC Name: 3-chloro-2-(chloromethyl)prop-1-ene
Synonyms: 3-chloro-2-(chloromethyl)prop-1-ene
Molecular Weight (g/mol): 124.99
Molecular Formula: C4H6Cl2
InChi Key: XJFZOSUFGSANIF-UHFFFAOYSA-N
SMILES: ClCC(=C)CCl
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)