missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-chloromethyl-1-propene 98.0+%, TCI America™

Catalog No. C15655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15655G 5 g
C156525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15655G Supplier TCI America Supplier No. C15655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1871-57-4
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000952
InChI Key XJFZOSUFGSANIF-UHFFFAOYSA-N
Synonym 3-chloro-2-chloromethyl prop-1-ene, 3-chloro-2-chloromethyl-1-propene, 1,1-bis chloromethyl ethylene, methallyl dichloride, 1-propene, 3-chloro-2-chloromethyl, 2-chloromethyl-3-chloropropene, 1,3-dichloro-2-methylenepropane, 3-chloro-2-chloromethyl propene, 2-methylene-1,3-dichloropropane, 2-methylenepropane-1,3-dichloride
PubChem CID 15859
IUPAC Name 3-chloro-2-(chloromethyl)prop-1-ene
SMILES ClCC(=C)CCl

Specifications

Melting Point -14°C
Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 2929
Formula Weight 124.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-chloromethyl-1-propene
RTECSNumber UC7400000
TSCA No
Recommended Storage Refrigerator