accessibility menu, dialog, popup

UserName

CH3OHO(E)

CAS RN 1866-39-3

IUPAC Name: (2E)-3-(4-methylphenyl)prop-2-enoic acid
Synonyms: 4-methyl-cinnamic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: RURHILYUWQEGOS-VOTSOKGWSA-N
SMILES: CC1=CC=C(\C=C\C(O)=O)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)