missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcinnamic Acid 98.0+%, TCI America™

Catalog No. M071525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M071525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M071525G Supplier TCI America Supplier No. M071525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1866-39-3
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00002697
InChI Key RURHILYUWQEGOS-VOTSOKGWSA-N
Synonym 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
PubChem CID 731767
IUPAC Name (E)-3-(4-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylcinnamic Acid