missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcinnamic acid, predominantly trans, 99%

Catalog No. AAA1333722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333722 100 g
AAA1333706 5 g
AAA1333714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1333722 Supplier Thermo Scientific Chemicals Supplier No. A1333722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1866-39-3 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1866-39-3
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00002697
InChI Key RURHILYUWQEGOS-VOTSOKGWSA-N
Synonym 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
PubChem CID 731767
IUPAC Name (E)-3-(4-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 196°C to 200°C
Quantity 100 g
Beilstein 1364327
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 4-Methylcinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-479-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only