accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNHCl

CAS RN 184719-80-0

IUPAC Name: tert-butyl pyrrolidine-2-carboxylate hydrochloride
Synonyms: tert-butyl pyrrolidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 207.7
Molecular Formula: C9H18ClNO2
InChi Key: IUUYANMOEMBTBV-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)C1CCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results