missing translation for 'onlineSavingsMsg'
Learn More

D-Proline tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. P17285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17285G Supplier TCI America Supplier No. P17285G
Only null left

Specifications

CAS 184719-80-0
Melting Point 94°C
Color White
Molecular Formula C9H18ClNO2
Quantity 5 g
MDL Number MFCD00153460
Synonym H-D-Pro-OtBu.HCl
InChI Key IUUYANMOEMBTBV-OGFXRTJISA-N
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl
IUPAC Name tert-butyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
Molecular Weight (g/mol) 207.698
PubChem CID 44629909
Formula Weight 207.70
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Proline tert-Butyl Ester Hydrochloride
Show More Show Less