accessibility menu, dialog, popup

UserName

CAS RN 18409-17-1

IUPAC Name: (2E)-oct-2-en-1-ol
Synonyms: (2E)-oct-2-en-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: AYQPVPFZWIQERS-VOTSOKGWSA-N
SMILES: CCCCC\C=C\CO
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)