missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octen-1-ol 95.0+%, TCI America™

Catalog No. O024710ML
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O024710ML 10 mL
O0247100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O024710ML Supplier TCI America Supplier No. O024710ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18409-17-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014057
InChI Key AYQPVPFZWIQERS-VOTSOKGWSA-N
Synonym trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol
PubChem CID 5318599
IUPAC Name (E)-oct-2-en-1-ol
SMILES CCCCCC=CCO

Specifications

Color Yellow
Boiling Point 92°C
Quantity 10 mL
Formula Weight 128.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Octen-1-ol
EINECSNumber (2)-2393
TSCA Yes