missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octen-1-ol, 97%

Catalog No. AAA1596022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1596022 100 g
AAA1596006 5 g
AAA1596014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1596022 Supplier Thermo Scientific Chemicals Supplier No. A1596022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18409-17-1 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18409-17-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014057
InChI Key AYQPVPFZWIQERS-VOTSOKGWSA-N
Synonym trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol
PubChem CID 5318599
IUPAC Name (E)-oct-2-en-1-ol
SMILES CCCCCC=CCO

Specifications

Density 0.85
Boiling Point 88°C (11 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.4465
Quantity 100 g
Beilstein 1720707
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material trans-2-Octen-1-ol
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 242-288-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only