accessibility menu, dialog, popup

UserName

CH3ClN

CAS RN 18368-63-3

IUPAC Name: 2-chloro-6-methylpyridine
Synonyms: 6-chloro-2-picoline
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: GXZDYRYYNXYPMQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=N1
Boiling Point 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)