missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyridine, 98%

Catalog No. AAB2017414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2017414 25 g
AAB2017406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2017414 Supplier Thermo Scientific Chemicals Supplier No. B2017414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18368-63-3 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1

Specifications

Density 1.167
Boiling Point 64°C to 68°C (10 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.5265
Quantity 25 g
UN Number UN2810
Beilstein 107187
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylpyridine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 242-241-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only