missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyridine 98.0+%, TCI America™

Catalog No. C16545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16545G 5 g
C165425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16545G Supplier TCI America Supplier No. C16545G

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 127.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-methylpyridine