accessibility menu, dialog, popup

UserName

BFFFFHOOH

CAS RN 182344-16-7

IUPAC Name: [4-fluoro-2-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-fluoro-2-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 207.92
Molecular Formula: C7H5BF4O2
InChi Key: SWUPLEAGZOKLNX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=C(F)C=C1)C(F)(F)F
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)