missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10521G Supplier TCI America Supplier No. F10521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 182344-16-7
Melting Point 186°C
Color White
Molecular Formula C7H5BF4O2
Quantity 1 g
MDL Number MFCD08062379
Synonym 4-fluoro-2-trifluoromethyl phenyl boronic acid, 4-fluoro-2-trifluoromethyl benzeneboronic acid, 4-fluoro-2-trifluoromethyl phenylboronic acid, 2-borono-5-fluorobenzotrifluoride, boronic acid, 4-fluoro-2-trifluoromethyl phenyl, akos brn-1027, acmc-1byq0, 4-fluoro-2-trifluoromethyl-phenylboronic acid, ksc495e5n
InChI Key SWUPLEAGZOKLNX-UHFFFAOYSA-N
SMILES OB(O)C1=C(C=C(F)C=C1)C(F)(F)F
IUPAC Name [4-fluoro-2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.92
PubChem CID 12109469
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less