accessibility menu, dialog, popup

UserName

CH3OOOHOH

CAS RN 1820579-68-7

IUPAC Name: 4-[(4-methoxyphenyl)methoxy]butane-1,3-diol
Synonyms: 4-[(4-methoxyphenyl)methoxy]butane-1,3-diol
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: GCVOAHFIFQYCKH-UHFFFAOYNA-N
SMILES: COC1=CC=C(COCC(O)CCO)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results