accessibility menu, dialog, popup

UserName

CAS RN 1797983-09-5

H3COOHNNNNNHCl

CAS RN 1797983-09-5

IUPAC Name: ethyl 3-{[2-(pyridin-2-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoate hydrochloride
Synonyms: ethyl 3-{[2-(pyridin-2-yl)-6-(1,2,4,5-tetrahydro-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoate hydrochloride
Molecular Weight (g/mol): 453.97
Molecular Formula: C24H28ClN5O2
InChi Key: TYXWLTBYINKVNT-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)CCNC1=CC(=NC(=N1)C1=CC=CC=N1)N1CCC2=CC=CC=C2CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results