CAS RN 17938-13-5
IUPAC Name:
trimethyl(2-{4-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)silane
Synonyms:
trimethyl(2-{4-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)silane
Molecular Weight (g/mol):
270.52
Molecular Formula:
C16H22Si2
InChi Key:
CMTMWEXUJQSPCA-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C
1
–
1
of
1
results
1
Promotions Available
1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%
1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%
CAS: 17938-13-5 Molecular Formula: C16H22Si2 Molecular Weight (g/mol): 270.522 MDL Number: MFCD00135433 InChI Key: CMTMWEXUJQSPCA-UHFFFAOYSA-N Synonym: 1,4-bis trimethylsilyl ethynyl benzene,1,4-bis trimethylsilylethynyl benzene,1,4-bis 2-trimethylsilyl ethynyl benzene,trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane,trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane,acmc-20ap2w,trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane,benzene,1,4-bis 2-trimethylsilyl ethynyl PubChem CID: 522492 IUPAC Name: trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane SMILES: C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C