missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%

Catalog No. AAL0970906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0970906 5 g
AAL0970903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0970906 Supplier Thermo Scientific Chemicals Supplier No. L0970906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17938-13-5 | C16H22Si2 | 270.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17938-13-5
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00135433
InChI Key CMTMWEXUJQSPCA-UHFFFAOYSA-N
Synonym 1,4-bis trimethylsilyl ethynyl benzene, 1,4-bis trimethylsilylethynyl benzene, 1,4-bis 2-trimethylsilyl ethynyl benzene, trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane, trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-20ap2w, trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane, benzene,1,4-bis 2-trimethylsilyl ethynyl
PubChem CID 522492
IUPAC Name trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Beilstein 2840618
Formula Weight 270.53
Percent Purity 98%
Chemical Name or Material 1,4-Bis[(trimethylsilyl)ethynyl]benzene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only