accessibility menu, dialog, popup

UserName

H3CSiH3CCH3SOO

CAS RN 17872-92-3

IUPAC Name: [(benzenesulfonyl)methyl]trimethylsilane
Synonyms: [(benzenesulfonyl)methyl]trimethylsilane
Molecular Weight (g/mol): 228.38
Molecular Formula: C10H16O2SSi
InChi Key: OEXJFEDAGJGOEP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results