accessibility menu, dialog, popup

UserName

CH3OHO(Z)(Z)(Z)

CAS RN 1783-84-2

IUPAC Name: (8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
Synonyms: dihomo-gamma-linolenic acid gamma-homolinolenic acid 8c,11c,14c-eicosatrienoic acid 8, 11, 14-eicosatrienoic acid star gla (gnc) 8c,11c,14c-eicosatriensaeure homo-gamma-linolenic acid homo-gamma-linolensaeure 8,11,14-eicosatrienoic acid bishomo-gamma-linolenic acid
Molecular Weight (g/mol): 306.49
Molecular Formula: C20H34O2
InChi Key: HOBAELRKJCKHQD-QNEBEIHSSA-N
SMILES: CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(O)=O
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results