missing translation for 'onlineSavingsMsg'
Learn More

all cis-8,11,14-Eicosatrienoic Acid 98.0+%, TCI America™

Catalog No. E064050MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E064050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E064050MG Supplier TCI America Supplier No. E064050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1783-84-2
Molecular Formula C20H34O2
Molecular Weight (g/mol) 306.49
MDL Number MFCD00065721
InChI Key HOBAELRKJCKHQD-QNEBEIHSSA-N
Synonym dihomo-gamma-linolenic acid, dgla, gamma-homolinolenic acid, 8,11,14-eicosatrienoic acid, homo-gamma-linolenic acid, cis-8,11,14-eicosatrienoic acid, bishomo-gamma-linolenic acid, z,z,z-8,11,14-eicosatrienoic acid, all-cis-8,11,14-eicosatrienoic acid, 8z,11z,14z-icosa-8,11,14-trienoic acid
PubChem CID 5280581
ChEBI CHEBI:53486
IUPAC Name (8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
SMILES CCCCCC=CCC=CCC=CCCCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 50 mg
Formula Weight 306.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material all cis-8,11,14-Eicosatrienoic Acid
EINECSNumber (2)-0609
RTECSNumber JX3880000
TSCA No
Recommended Storage Freezer (-20 °C)