accessibility menu, dialog, popup

UserName

CAS RN 17768-41-1

IUPAC Name: (adamantan-1-yl)methanaminium
Synonyms: adamantan-1-ylmethanaminium
Molecular Weight (g/mol): 166.29
Molecular Formula: C11H20N
InChi Key: XSOHXMFFSKTSIT-UHFFFAOYSA-O
SMILES: [NH3+]CC12CC3CC(CC(C3)C1)C2
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)