missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethylamine 98.0+%, TCI America™

Catalog No. A07215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07215G 5 g
A07211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07215G Supplier TCI America Supplier No. A07215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17768-41-1
Molecular Formula C11H20N
Molecular Weight (g/mol) 166.29
MDL Number MFCD00074750
InChI Key XSOHXMFFSKTSIT-UHFFFAOYSA-O
Synonym 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
PubChem CID 86625
IUPAC Name (adamantan-1-yl)methanaminium
SMILES [NH3+]CC12CC3CC(CC(C3)C1)C2

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 165.28
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Adamantanemethylamine