missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethylamine, 98%, Thermo Scientific™

Catalog No. p-3777176
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177420010 1g Glass bottle
AC177420050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177420010 Supplier Thermo Scientific Chemicals Supplier No. 177420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 17768-41-1
Molecular Formula C11H20N
Molecular Weight (g/mol) 166.29
MDL Number MFCD00074750
InChI Key XSOHXMFFSKTSIT-UHFFFAOYSA-O
Synonym 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
PubChem CID 86625
IUPAC Name 1-adamantylmethanamine
SMILES [NH3+]CC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9300g/mL
Boiling Point 83.0°C to 85.0°C (0.3mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5127 to 1.5147
Quantity 1g
Specific Gravity 0.93
Formula Weight 165.28
Percent Purity 98%
Chemical Name or Material 1-Adamantanemethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 241-752-1

RUO – Research Use Only