accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 17746-05-3

IUPAC Name: undecanoyl chloride
Synonyms: undecanoyl chloride
Molecular Weight (g/mol): 204.74
Molecular Formula: C11H21ClO
InChi Key: JUKPJGZUFHCZQI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(Cl)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)