missing translation for 'onlineSavingsMsg'
Learn More

n-Undecanoyl chloride, 98%

Catalog No. AAH56380ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56380ME 500 mg
AAH5638003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56380ME Supplier Thermo Scientific Chemicals Supplier No. H56380ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17746-05-3 | C11H21ClO | 204.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17746-05-3
Molecular Formula C11H21ClO
Molecular Weight (g/mol) 204.738
MDL Number MFCD00000739
InChI Key JUKPJGZUFHCZQI-UHFFFAOYSA-N
Synonym n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
PubChem CID 87287
IUPAC Name undecanoyl chloride
SMILES CCCCCCCCCCC(=O)Cl

Specifications

Boiling Point 135°C to 136°C
Flash Point 113°C (235°F)
Refractive Index 1.443
Linear Formula CH3(CH2)9COCl
Quantity 500 mg
UN Number UN3265
Beilstein 1759226
Sensitivity Moisture sensitive
Formula Weight 204.74
Percent Purity 98%
Chemical Name or Material n-Undecanoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 241-741-1
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only