missing translation for 'onlineSavingsMsg'
Learn More

Undecanoyl Chloride 97.0+%, TCI America™

Catalog No. U002625ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U002625ML 25 mL
U002610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U002625ML Supplier TCI America Supplier No. U002625ML

Chemical Identifiers

CAS 17746-05-3
Molecular Formula C11H21ClO
Molecular Weight (g/mol) 204.738
MDL Number MFCD00000739
InChI Key JUKPJGZUFHCZQI-UHFFFAOYSA-N
Synonym n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
PubChem CID 87287
IUPAC Name undecanoyl chloride
SMILES CCCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 3265
Formula Weight 204.74
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Undecanoyl Chloride