missing translation for 'onlineSavingsMsg'
Learn More

8-Bromooctanoic acid, 95%

Catalog No. AAB2557518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2557518 50 g
AAB2557509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2557518 Supplier Thermo Scientific Chemicals Supplier No. B2557518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17696-11-6 | C8H15BrO2 | 223.11 g/mol

8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4'-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4′-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases.

Chemical Identifiers

CAS 17696-11-6
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00004430
InChI Key BKJFDZSBZWHRNH-UHFFFAOYSA-N
Synonym 8-bromocaprylic acid, 8-bromo octanoic acid, octanoic acid, 8-bromo, 8-bromo-n-octanoic acid, omega-bromocaprylic acid, 8-bromo-octanoic acid, octanoicacid, 8-bromo, acmc-1bo24, 8-bromooctanoic acid, ksc180g8h
PubChem CID 548275
IUPAC Name 8-bromooctanoic acid
SMILES C(CCCC(=O)O)CCCBr

Specifications

Melting Point 37°C to 41°C
Boiling Point 147°C to 150°C (2 mmHg)
Flash Point >110°C (230°F)
Linear Formula BrCH2(CH2)6CO2H
Quantity 50 g
UN Number UN1759
Solubility Information Slightly soluble in water.
Formula Weight 223.11
Percent Purity 97%
Chemical Name or Material 8-Bromooctanoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only