accessibility menu, dialog, popup

UserName

CH3OCH3OCH3OH3COCH3OH3CONNEu(E/Z)(E/Z)(E/Z)

CAS RN 17568-09-1

IUPAC Name: europium(3+) 1,10-phenanthroline tris(4-oxopent-2-en-2-olate)
Synonyms: europium(3+) phen tris(4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 629.5
Molecular Formula: C27H29EuN2O6
InChi Key: UDXZANKTSSCELS-UHFFFAOYSA-K
SMILES: [Eu+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results