missing translation for 'onlineSavingsMsg'
Learn More

Tris(acetylacetonato)(1,10-phenanthroline)europium(III) 98.0+%, TCI America™

Catalog No. T3206200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3206200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3206200MG Supplier TCI America Supplier No. T3206200MG
Only null left

Chemical Identifiers

CAS 17568-09-1
Molecular Formula C27H29EuN2O6
Molecular Weight (g/mol) 629.501
InChI Key UDXZANKTSSCELS-XUHIWKAKSA-K
PubChem CID 91972102
IUPAC Name europium(3+);(Z)-4-oxopent-2-en-2-olate;1,10-phenanthroline
SMILES CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu+3]

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 200 mg
Formula Weight 629.50
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(acetylacetonato)(1,10-phenanthroline)europium(III)