accessibility menu, dialog, popup

UserName

BNNNNNNCH3CH3CH3H3CH3CH3CHK

CAS RN 17567-17-8

IUPAC Name: potassium tris(3,5-dimethyl-1H-pyrazol-1-yl)boranuide
Synonyms: potassium tris(3,5-dimethylpyrazol-1-yl)boranuide
Molecular Weight (g/mol): 336.29
Molecular Formula: C15H22BKN6
InChi Key: NTWZGFNSHCFHIJ-UHFFFAOYSA-N
SMILES: [K+].CC1=NN([BH-](N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)