accessibility menu, dialog, popup

UserName

CAS RN 17515-77-4

OFFFBr

CAS RN 17515-77-4

IUPAC Name: 2-(bromomethyl)-5-(trifluoromethyl)furan
Synonyms: 2-(bromomethyl)-5-(trifluoromethyl)furan
Molecular Weight (g/mol): 229
Molecular Formula: C6H4BrF3O
InChi Key: YNHVBNGRNNVEMD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CBr)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results