accessibility menu, dialog, popup

UserName

OH2NO

CAS RN 175136-34-2

IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
Synonyms: 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: FVLCICVRAPEYNX-UHFFFAOYSA-N
SMILES: NC1=CC2=C(OCCCO2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)