accessibility menu, dialog, popup

UserName

NFHO

CAS RN 174669-74-0

IUPAC Name: 2-fluoropyridin-3-ol
Synonyms: 2-fluoropyridin-3-ol
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: UEQRKEWMEMJXQO-UHFFFAOYSA-N
SMILES: OC1=C(F)N=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results