missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-hydroxypyridine 98.0+%, TCI America™

Catalog No. F08865G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08865G 5 g
F08861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08865G Supplier TCI America Supplier No. F08865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 174669-74-0 | C5H4FNO | 113.09 g/mol

Chemical Identifiers

CAS 174669-74-0
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.09
MDL Number MFCD04112569
InChI Key UEQRKEWMEMJXQO-UHFFFAOYSA-N
Synonym 2-fluoro-3-hydroxypyridine, 2-fluoro-3-pyridinol, 2-fluoropyridine-3-ol, 3-pyridinol, 2-fluoro, 2-fluoro-3-hydroxy pyridine, pubchem6290, acmc-209e8o, 3-hydroxy-2-fluoropyridine, 3-pyridinol, 2-fluoro-9ci
PubChem CID 2762785
IUPAC Name 2-fluoropyridin-3-ol
SMILES OC1=C(F)N=CC=C1

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 5 g
Formula Weight 113.09
Percent Purity ≥98.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-hydroxypyridine