accessibility menu, dialog, popup

UserName

CAS RN 17440-83-4

IUPAC Name: hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide dihydrate chloride
Synonyms: hydrogen amiloride dihydrate chloride
Molecular Weight (g/mol): 302.12
Molecular Formula: C6H13Cl2N7O3
InChi Key: LTKVFMLMEYCWMK-UHFFFAOYSA-N
SMILES: [H+].O.O.[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results