accessibility menu, dialog, popup

UserName

H3COOCH2Cl

CAS RN 17435-77-7

IUPAC Name: ethyl 2-(chloromethyl)prop-2-enoate
Synonyms: ethyl 2-(chloromethyl)prop-2-enoate
Molecular Weight (g/mol): 148.59
Molecular Formula: C6H9ClO2
InChi Key: KPXRGIVPSXFJEX-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=C)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results