accessibility menu, dialog, popup

UserName

CH3CH3CH3OH3CH3C

CAS RN 17432-38-1

IUPAC Name: 2,3,4,5,6-pentamethylbenzaldehyde
Synonyms: 2,3,4,5,6-pentamethylbenzaldehyde
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: RWOZGGOKRKSHKN-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C=O)C(C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results