accessibility menu, dialog, popup

UserName

ClOOClClCl

CAS RN 17341-93-4

IUPAC Name: 2,2,2-trichloroethyl carbonochloridate
Synonyms: 2,2,2-trichloroethyl carbonochloridate
Molecular Weight (g/mol): 211.85
Molecular Formula: C3H2Cl4O2
InChi Key: LJCZNYWLQZZIOS-UHFFFAOYSA-N
SMILES: ClC(=O)OCC(Cl)(Cl)Cl
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)