accessibility menu, dialog, popup

UserName

CAS RN 17302-82-8

IUPAC Name: ethyl 3,5-dichloro-4-hydroxybenzoate
Synonyms: ethyl 3,5-dichloro-4-hydroxybenzoate
Molecular Weight (g/mol): 235.06
Molecular Formula: C9H8Cl2O3
InChi Key: WMKNGSJJEMFQOT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(Cl)=C(O)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results