accessibility menu, dialog, popup

UserName

H3COOOONNONOOOOH3COOOOCH3OOONOOOOOCH3NOOOOCH3OOOOCH3

CAS RN 172890-84-5

IUPAC Name: (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}[5-(2-{5-[3-(4-{2-[(acetyloxy)methoxy]-2-oxoethyl}piperazin-1-yl)-3-oxopropyl]-2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]phenoxy}ethoxy)-2-(5-{[(acetyloxy)methoxy]carbonyl}-1,3-oxazol-2-yl)-1-benzofuran-6-yl]amino)acetate
Synonyms: (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}[5-(2-{5-[3-(4-{2-[(acetyloxy)methoxy]-2-oxoethyl}piperazin-1-yl)-3-oxopropyl]-2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]phenoxy}ethoxy)-2-(5-{[(acetyloxy)methoxy]carbonyl}-1,3-oxazol-2-yl)-1-benzofuran-6-yl]amino)acetate
Molecular Weight (g/mol): 1258.12
Molecular Formula: C55H63N5O29
InChi Key: CGAZNAYDEAJZHQ-UHFFFAOYSA-N
SMILES: CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)C1=C(OCCOC2=C(C=C3OC(=CC3=C2)C2=NC=C(O2)C(=O)OCOC(C)=O)N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C=C(CCC(=O)N2CCN(CC(=O)OCOC(C)=O)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results